Learn More
Medchemexpress LLC Complement C5-IN-1 | 2365402-67-9 | 99.1% | 10 MG
SDP
Catalog No. 5000341207
Encompass_Preferred
Complement C5-IN-1 (Compound 7) is a selective allosteric inhibitor of complement component protein C5. It prevents C5 from being cleaved by C5 convertase, inhibiting the cleavage of C5 into C5a and C5b, and thus blocking the formation of membrane attack complex (MAC). It can be used to study diseases related to complement overactivation, such as paroxysmal nocturnal hemoglobinuria (PNH) and atypical hemolytic uremic syndrome (aHUS).
  • Selective allosteric inhibitor of complement component protein C5
  • Prevents C5 cleavage by C5 convertase
  • Inhibits C5a and C5b formation
  • Blocks membrane attack complex (MAC) formation
  • Useful for studying diseases related to complement overactivation
Catalog No. 50-003-41207 Supplier Medchemexpress LLC Supplier No. HY12834210MG
Complement C5-IN-1 (Compound 7) is a selective allosteric inhibitor of complement component protein C5. It prevents C5 from being cleaved by C5 convertase, inhibiting the cleavage of C5 into C5a and C5b, and thus blocking the formation of membrane attack complex (MAC). It can be used to study diseases related to complement overactivation, such as paroxysmal nocturnal hemoglobinuria (PNH) and atypical hemolytic uremic syndrome (aHUS).
  • Selective allosteric inhibitor of complement component protein C5
  • Prevents C5 cleavage by C5 convertase
  • Inhibits C5a and C5b formation
  • Blocks membrane attack complex (MAC) formation
  • Useful for studying diseases related to complement overactivation