Learn More
Cusabio Technology LLC complement component 2
Catalog No. 5000051127
Encompass_Preferred
The Mouse Complement C2 (C2) ELISA Kit is designed to quantitatively measure C2 levels in serum plasma and tissue homogenates using a competitive assay format This kit detects C2 with a sensitivity of 18 75 ng/mL and a dynamic detection range of 18 75-300 ng/mL ensuring accurate quantification across physiological and experimental concentrations The assay requires a sample volume of 50-100 L and features a streamlined protocol with an incubation time of 1-5 hours optimized for efficient workflow integration Detection is performed at 450 nm providing reliable absorbance measurements for precise data analysis Ideal for research applications this kit offers robust performance in quantifying complement C2 levels supporting studies in immunology inflammation and related fields Its competitive binding methodology ensures specificity and reproducibility making it suitable for both routine laboratory use and specialized investigative projects
Catalog No. 50-000-51127 Supplier Cusabio Technology LLC Supplier No. CSBEL003658MO
The Mouse Complement C2 (C2) ELISA Kit is designed to quantitatively measure C2 levels in serum plasma and tissue homogenates using a competitive assay format This kit detects C2 with a sensitivity of 18 75 ng/mL and a dynamic detection range of 18 75-300 ng/mL ensuring accurate quantification across physiological and experimental concentrations The assay requires a sample volume of 50-100 L and features a streamlined protocol with an incubation time of 1-5 hours optimized for efficient workflow integration Detection is performed at 450 nm providing reliable absorbance measurements for precise data analysis Ideal for research applications this kit offers robust performance in quantifying complement C2 levels supporting studies in immunology inflammation and related fields Its competitive binding methodology ensures specificity and reproducibility making it suitable for both routine laboratory use and specialized investigative projects