Learn More
Biomatik Corporation Complement Component 4C (C4C) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU30135
Encompass_Preferred
Complement Component 4C (C4C) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Complement Component 4c (C4c). The Complement Component 4C (C4C) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Human (Homo sapiens). Applications: WB, IHC, ICC, IP. Target Synonyms: Complement Component 4c (C4c). Accession Number: P0C0L5. Gene ID: 100293534. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6552