Learn More
Art Robbins Instruments Complement cups, 100 per bag
SDP
Catalog No. 501530633
Encompass_Preferred
This item has a minimum qty of 2 per supplier requirements.
Complement cups, 10mL volume, made form non-cytotoxic, non-wettable virgin polypropylene, autoclavable, 100/BG
Catalog No. 50-153-0633 Supplier Art Robbins Instruments Supplier No. 103600000
This item has a minimum qty of 2 per supplier requirements.
Complement cups, 10mL volume, made form non-cytotoxic, non-wettable virgin polypropylene, autoclavable, 100/BG