Learn More
Alpco Ltd Canine Complement Factor 3 ELISA
SDP
Supplier:  Alpco Ltd 41CO3CAE01
Encompass
The Canine Complement Factor 3 ELISA is a highly sensitive two-site enzyme linked immunoassay (ELISA) for measuring C3 in biological samples of dogs. Research Use Only. Not for Use in Diagnostic Procedures.
Catalog No. 50-751-3573