Learn More
Medchemexpress LLC Complement Factor D/ 50ug | 50ug
SDP
Catalog No. 5000187481
Encompass_Preferred
Complement Factor D/Adipsin Protein cleaves factor B in complex with factor C3b activating the C3bbb complex to form the C3 convertase in the alternate pathway analogous to C1s in the classical pathway Complement Factor D/Adipsin Protein Mouse (HEK293 Fc) is the recombinant mouse-derived Complement Factor D/Adipsin protein expressed by HEK293 with C-hFc labeled tag
Catalog No. 50-001-87481 Supplier Medchemexpress LLC Supplier No. HYP7827350UG
Complement Factor D/Adipsin Protein cleaves factor B in complex with factor C3b activating the C3bbb complex to form the C3 convertase in the alternate pathway analogous to C1s in the classical pathway Complement Factor D/Adipsin Protein Mouse (HEK293 Fc) is the recombinant mouse-derived Complement Factor D/Adipsin protein expressed by HEK293 with C-hFc labeled tag