Learn More
Medchemexpress LLC Complement Factor D/ 50ug | 50ug
SDP
Catalog No. 5000194142
Encompass_Preferred
Complement factor D also known as Adipsin plays a crucial role in the alternative pathway of the complement system and when complexed with factor C3b it cleaves factor B This cleavage activates the C3bbb complex converting it to the C3 convertase of the alternative pathway Complement Factor D/Adipsin Protein Human (HEK293 His Solution) is the recombinant human-derived Complement Factor D/Adipsin protein expressed by HEK293 with C-10 His labeled tag
Catalog No. 50-001-94142 Supplier Medchemexpress LLC Supplier No. HYP7861A50UG
Complement factor D also known as Adipsin plays a crucial role in the alternative pathway of the complement system and when complexed with factor C3b it cleaves factor B This cleavage activates the C3bbb complex converting it to the C3 convertase of the alternative pathway Complement Factor D/Adipsin Protein Human (HEK293 His Solution) is the recombinant human-derived Complement Factor D/Adipsin protein expressed by HEK293 with C-10 His labeled tag