Learn More
Medchemexpress LLC Complement Factor D/ 50ug
SDP
Catalog No. 5000195527
Encompass_Preferred
Recombinant rat Complement Factor D (Adipsin) expressed in Pichia pastoris with an N-terminal 6xHis tag. Supplied as a lyophilized powder formulated in PBS with 6% trehalose, the protein is glycosylated, >95% pure by reducing SDS-PAGE, and has low endotoxin (<1 EU/μg). Intended for research on the alternative complement pathway; store at -20°C and follow reconstitution guidance (do not reconstitute below 100 μg/mL).
  • Recombinant protein expressed in Pichia pastoris.
  • N-terminal 6xHis tag for purification or detection.
  • Lyophilized formulation in PBS with 6% trehalose for stability.
  • High purity greater than 95% by reducing SDS-PAGE.
  • Low endotoxin level below 1 EU/μg.
  • Glycosylated; migrates ~43 kDa under reducing conditions.
  • Stable when stored at -20°C; aliquot after reconstitution for long-term storage.
Catalog No. 50-001-95527 Supplier Medchemexpress LLC Supplier No. HYP7172650UG
Recombinant rat Complement Factor D (Adipsin) expressed in Pichia pastoris with an N-terminal 6xHis tag. Supplied as a lyophilized powder formulated in PBS with 6% trehalose, the protein is glycosylated, >95% pure by reducing SDS-PAGE, and has low endotoxin (<1 EU/μg). Intended for research on the alternative complement pathway; store at -20°C and follow reconstitution guidance (do not reconstitute below 100 μg/mL).
  • Recombinant protein expressed in Pichia pastoris.
  • N-terminal 6xHis tag for purification or detection.
  • Lyophilized formulation in PBS with 6% trehalose for stability.
  • High purity greater than 95% by reducing SDS-PAGE.
  • Low endotoxin level below 1 EU/μg.
  • Glycosylated; migrates ~43 kDa under reducing conditions.
  • Stable when stored at -20°C; aliquot after reconstitution for long-term storage.