Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Complement Factor D/ 5ug
SDP

Catalog No. 5000195927
Encompass_Preferred

Recombinant rat Complement Factor D (Adipsin) produced in Pichia pastoris and provided as a purified, His-tagged protein for research applications involving the alternative complement pathway. The protein is supplied in microgram quantities and is suitable for proteolytic activity assays, antibody binding studies, and other in vitro experiments; store frozen and aliquot to preserve activity.

  • Recombinant rat-derived protein for alternative complement pathway studies.
  • Expressed in Pichia pastoris with an N-terminal 6xHis tag for easy purification.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Apparent molecular weight of approximately 43 kDa, consistent with glycosylation.
  • Stable when stored at -20°C; aliquot and store at -80°C for extended storage.

Catalog No. 50-001-95927 Supplier Medchemexpress LLC Supplier No. HYP717265UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant rat Complement Factor D (Adipsin) produced in Pichia pastoris and provided as a purified, His-tagged protein for research applications involving the alternative complement pathway. The protein is supplied in microgram quantities and is suitable for proteolytic activity assays, antibody binding studies, and other in vitro experiments; store frozen and aliquot to preserve activity.

  • Recombinant rat-derived protein for alternative complement pathway studies.
  • Expressed in Pichia pastoris with an N-terminal 6xHis tag for easy purification.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Apparent molecular weight of approximately 43 kDa, consistent with glycosylation.
  • Stable when stored at -20°C; aliquot and store at -80°C for extended storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.