Learn More
Medchemexpress LLC Complement factor D-IN-1 | 1386455-76-0 | 98.3% | 483.32 g/mol | C21H19BrN6O3 | 1 ML
SDP
Catalog No. 5000247442
Encompass_Preferred
Complement factor D-IN-1 is a small-molecule, reversible inhibitor of complement factor D used as a biochemical probe in complement pathway and immunology research. It is supplied as a 10 mM solution in DMSO (1 mL) and is also available as solid quantities; the compound is provided with documentation to support research use.
Catalog No. 50-002-47442 Supplier Medchemexpress LLC Supplier No. HY10203410MM 1ML
Complement factor D-IN-1 is a small-molecule, reversible inhibitor of complement factor D used as a biochemical probe in complement pathway and immunology research. It is supplied as a 10 mM solution in DMSO (1 mL) and is also available as solid quantities; the compound is provided with documentation to support research use.