Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Complement factor D-IN-1 | 1386455-76-0 | 98.9% | 483.32 g·mol⁻1 | C21H19BrN6O3 | 10 MG
SDP

Catalog No. 5000246655
Encompass_Preferred

Complement factor D-IN-1 is a potent, selective, small-molecule reversible inhibitor of complement factor D intended for research into complement system biology and complement-mediated inflammation. It has been characterized in biochemical assays and is supplied as solid or solution for laboratory studies.

  • Potent inhibitory activity: IC50 = 0.006 μM (FD thioesterolytic assay) and IC50 = 0.05 μM (MAC deposition assay).
  • High reported purity (~98.9%); consult lot-specific COA for exact value.
  • Supplied as solid or as DMSO solution for flexible experimental workflows.
  • Stable storage: powder at -20°C (long term) or 4°C; in solvent, -80°C recommended.
  • Molecular weight 483.32 g·mol⁻1 and formula C21H19BrN6O3 for analytical reference.
  • For research use only; not for human or clinical applications.

Catalog No. 50-002-46655 Supplier Medchemexpress LLC Supplier No. HY10203410MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Complement factor D-IN-1 is a potent, selective, small-molecule reversible inhibitor of complement factor D intended for research into complement system biology and complement-mediated inflammation. It has been characterized in biochemical assays and is supplied as solid or solution for laboratory studies.

  • Potent inhibitory activity: IC50 = 0.006 μM (FD thioesterolytic assay) and IC50 = 0.05 μM (MAC deposition assay).
  • High reported purity (~98.9%); consult lot-specific COA for exact value.
  • Supplied as solid or as DMSO solution for flexible experimental workflows.
  • Stable storage: powder at -20°C (long term) or 4°C; in solvent, -80°C recommended.
  • Molecular weight 483.32 g·mol⁻1 and formula C21H19BrN6O3 for analytical reference.
  • For research use only; not for human or clinical applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.