Learn More
Medchemexpress LLC HY-102034 5mg Medchemexpress, Complement factor D-IN-1 CAS:1386455-76-0 Purity:>98%
SDP
Catalog No. 501967962
Encompass_Preferred
Medchemexpress, HY-102034 5mg Complement factor D-IN-1 CAS:1386455-76-0 Complement factor D-IN-1 is a potent and selective small-molecule reversible factor d inhibitor, with IC50s of 0.006 and 0.05 μM in FD Thioesterolytic Fluorescent Assay and a MAC Deposition Assay, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7962 Supplier Medchemexpress LLC Supplier No. HY1020345MG
Medchemexpress, HY-102034 5mg Complement factor D-IN-1 CAS:1386455-76-0 Complement factor D-IN-1 is a potent and selective small-molecule reversible factor d inhibitor, with IC50s of 0.006 and 0.05 μM in FD Thioesterolytic Fluorescent Assay and a MAC Deposition Assay, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.