Learn More
Medchemexpress LLC Complement factor D-IN-2 | 1903742-34-6 | 99.33% | 574.43 | 25 MG
SDP
Catalog No. 5000352428
Encompass_Preferred
Complement factor D-IN-2 is an inhibitor of complement factor D, extracted from patent WO2015130838A1, compound 190. It targets factor D and inhibits the complement cascade at an early and essential point in the alternative complement pathway, and can be used for the research of autoimmune diseases.
  • Reduces the excessive activation of complement factor D.
  • Targets factor D and inhibits the complement cascade.
  • Suitable for research of autoimmune diseases.
  • For research use only.
Catalog No. 50-003-52428 Supplier Medchemexpress LLC Supplier No. 25MGHY138281
Complement factor D-IN-2 is an inhibitor of complement factor D, extracted from patent WO2015130838A1, compound 190. It targets factor D and inhibits the complement cascade at an early and essential point in the alternative complement pathway, and can be used for the research of autoimmune diseases.
  • Reduces the excessive activation of complement factor D.
  • Targets factor D and inhibits the complement cascade.
  • Suitable for research of autoimmune diseases.
  • For research use only.