Learn More
Cusabio Technology LLC complement fragment 3b C3b
Catalog No. 5000049480
Encompass_Preferred
This kit detects C3b in mouse serum plasma and tissue homogenates with high sensitivity and precision The assay has a detection range of 0 235 ng/mL to 15 ng/mL enabling accurate quantification of both low and high concentration samples With a sensitivity of 0 059 ng/mL it reliably identifies even minimal levels of C3b making it suitable for applications requiring precise measurement in limited sample volumes (50-100 L) Utilizing a sandwich ELISA format the kit provides robust and specific results within 1 to 5 hours The absorbance is measured at 450 nm ensuring clear and consistent readouts for quantitative analysis This optimized workflow balances speed and accuracy catering to diverse experimental needs in complement system research
Catalog No. 50-000-49480 Supplier Cusabio Technology LLC Supplier No. CSBEQ027757MO
This kit detects C3b in mouse serum plasma and tissue homogenates with high sensitivity and precision The assay has a detection range of 0 235 ng/mL to 15 ng/mL enabling accurate quantification of both low and high concentration samples With a sensitivity of 0 059 ng/mL it reliably identifies even minimal levels of C3b making it suitable for applications requiring precise measurement in limited sample volumes (50-100 L) Utilizing a sandwich ELISA format the kit provides robust and specific results within 1 to 5 hours The absorbance is measured at 450 nm ensuring clear and consistent readouts for quantitative analysis This optimized workflow balances speed and accuracy catering to diverse experimental needs in complement system research