Learn More
Cusabio Technology LLC Complement fragment 4b C4b
Catalog No. 5000049321
Encompass_Preferred
This kit detects C4b in human serum and plasma samples using a quantitative sandwich ELISA method It offers a detection range of 15 6 ng/mL to 1000 ng/mL with a sensitivity of 3 9 ng/mL ensuring precise measurement of C4b concentrations The assay requires 50-100 L of sample and can be completed within 1-5 hours Absorbance is measured at 450 nm to quantify results making it suitable for research or clinical applications requiring reliable assessment of complement fragment 4b levels The standardized protocol and optimized reagents ensure consistent performance for both low and high concentration samples
Catalog No. 50-000-49321 Supplier Cusabio Technology LLC Supplier No. CSBE09302H
This kit detects C4b in human serum and plasma samples using a quantitative sandwich ELISA method It offers a detection range of 15 6 ng/mL to 1000 ng/mL with a sensitivity of 3 9 ng/mL ensuring precise measurement of C4b concentrations The assay requires 50-100 L of sample and can be completed within 1-5 hours Absorbance is measured at 450 nm to quantify results making it suitable for research or clinical applications requiring reliable assessment of complement fragment 4b levels The standardized protocol and optimized reagents ensure consistent performance for both low and high concentration samples