Learn More
IBL America Complement System Classical Pathway
Supplier:  IBL America COMPL CP310 RUO
Encompass_Preferred
The Wieslab COMPL CP310 is an enzyme immunoassay (ELISA) for the determination of functional classical complement pathway in human serum. For research use only, not for use in diagnostic procedures.
Catalog No. 50-136-6591