Learn More
IBL America Complement System Alternative Pathway
Supplier:  IBL America COMPL AP330 RUO
Encompass_Preferred
The Wieslab COMPL AP330 kit is an enzyme immunoassay (ELISA) for the determination of functional alternative complement pathway in human serum. For research use only, not for use in diagnostic procedures.
Catalog No. 50-136-6590