Learn More
IBL America Complement System Screen
Catalog No. NC1372880
Encompass_Preferred
The Wieslab Complement system Screen kit is an enzyme immunoassay for the determination of functional classical, MBL and alternative complement pathways in human serum. For research use only, not for use in diagnostic procedures.
The complement system plays an essential role in chronic, autoimmune and infectious disease. There are three pathways of complement activation, namely the classical, the alternative and the recently discovered MBL pathway.
Impaired complement activity causes humans to become susceptible to repetitive fulminant or severe infections and may contribute to development of autoimmune disease. Inappropriate activation of complement contributes to chronic inflammation and tissue injury. For research use only, not for use in diagnostic procedures.
Catalog No. NC1372880 Supplier IBL America Supplier No. COMPL 300 RUO
The Wieslab Complement system Screen kit is an enzyme immunoassay for the determination of functional classical, MBL and alternative complement pathways in human serum. For research use only, not for use in diagnostic procedures.
The complement system plays an essential role in chronic, autoimmune and infectious disease. There are three pathways of complement activation, namely the classical, the alternative and the recently discovered MBL pathway.
Impaired complement activity causes humans to become susceptible to repetitive fulminant or severe infections and may contribute to development of autoimmune disease. Inappropriate activation of complement contributes to chronic inflammation and tissue injury. For research use only, not for use in diagnostic procedures.