Learn More
ACROBiosystems ACROBiosystems Rabbit Complement C5 Protein, His Tag
Catalog No. 501956499
Encompass_Preferred
Recombinant Protein;1MG;Rabbit Complement C5, His Tag (CO5-R52H4) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Ala 1678 (Accession # XP_008271599.1).;This protein carries a polyhistidine tag at the C-terminus. The mature form of Complement C5 is a disulfide-linked heterodimer composed of proteolytically cleaved α and β chain. During cultivation, Complement C5 will be partially processed into α and β chain and has a calculated MW of 73.8 kDa (β chain) and 111.9 kDa (α chain) respectively. The protein migrates as 80 kDa (β chain), 120 kDa (α chain) and 200 kDa (α & β chain) under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-195-6499 Supplier ACROBiosystems Supplier No. CO5R52H41MG
Recombinant Protein;1MG;Rabbit Complement C5, His Tag (CO5-R52H4) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Ala 1678 (Accession # XP_008271599.1).;This protein carries a polyhistidine tag at the C-terminus. The mature form of Complement C5 is a disulfide-linked heterodimer composed of proteolytically cleaved α and β chain. During cultivation, Complement C5 will be partially processed into α and β chain and has a calculated MW of 73.8 kDa (β chain) and 111.9 kDa (α chain) respectively. The protein migrates as 80 kDa (β chain), 120 kDa (α chain) and 200 kDa (α & β chain) under reducing (R) condition (SDS-PAGE) due to glycosylation.