Learn More
ACROBiosystems ACROBiosystems Human Complement C5 (w917s) Protein, His Tag
Catalog No. 501956696
Encompass_Preferred
Recombinant Protein;100UG;Human Complement C5 (w917s), His Tag (CO5-H52H7) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Cys 1676 (Accession # P01031-1 (W917S)).;This protein carries a polyhistidine tag at the C-terminus. The mature form of Complement C5 is a disulfide-linked heterodimer composed of proteolytically cleaved α and β chain. Each α and β chain has a calculated MW of 73.9 kDa (β chain) and 114.3 kDa (α chain). The protein migrates as 80 kDa (β chain) and 110 kDa (α chain) under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human Complement C5 (w917s), His Tag (Cat. No. CO5-H52H7) at 2 μg/mL (100 μL/well) can bind Anti-C5a MAb (Human IgG1) with a linear range of 0.2-6 ng/mL (QC tested).
Catalog No. 50-195-6696 Supplier ACROBiosystems Supplier No. CO5H52H7100UG
Recombinant Protein;100UG;Human Complement C5 (w917s), His Tag (CO5-H52H7) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Cys 1676 (Accession # P01031-1 (W917S)).;This protein carries a polyhistidine tag at the C-terminus. The mature form of Complement C5 is a disulfide-linked heterodimer composed of proteolytically cleaved α and β chain. Each α and β chain has a calculated MW of 73.9 kDa (β chain) and 114.3 kDa (α chain). The protein migrates as 80 kDa (β chain) and 110 kDa (α chain) under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human Complement C5 (w917s), His Tag (Cat. No. CO5-H52H7) at 2 μg/mL (100 μL/well) can bind Anti-C5a MAb (Human IgG1) with a linear range of 0.2-6 ng/mL (QC tested).