Learn More
ACROBiosystems ACROBiosystems Mouse Complement C5 Protein, His Tag
Supplier:  ACROBiosystems CO5M52H41MG
Encompass_Preferred
Recombinant Protein;1MG;Mouse Complement C5, His Tag (CO5-M52H4) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Glu 1680 (Accession # P06684-1).;This protein carries a polyhistidine tag at the C-terminus. The mature form of Complement C5 is a disulfide-linked heterodimer composed of proteolytically cleaved α and β chain. Each α and β chain has a calculated MW of 80.0 kDa (β chain) and 110.0 kDa (α chain). The protein migrates as 66 kDa (β chain) and 100 kDa (α chain) under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Mouse Complement C5, His Tag (Cat. No. CO5-M52H4) at 2 μg/mL (100 μL/well) can bind Anti-C5a (Human IgG1) with a linear range of 0.2-6 ng/mL (QC tested).
Catalog No. 50-195-6912