Learn More
ACROBiosystems ACROBiosystems Rat Complement C5 Protein, His Tag
Catalog No. 501956905
Encompass_Preferred
Recombinant Protein;100UG;Rat Complement C5, His Tag (CO5-R52H5) is expressed from human 293 cells (HEK293). It contains AA Gln 24 - Ala 1688 (Accession # XP_001079130.2).;This protein carries a polyhistidine tag at the C-terminus.The mature form of Complement C5 is a disulfide-linked heterodimer composed of proteolytically cleaved α and β chain. Each α and β chain has a calculated MW of 74.1 kDa (β chain) and 114.8 kDa (α chain). The protein migrates as 70 kDa (β chain) and 110 kDa (α chain) under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Rat Complement C5, His Tag (Cat. No. CO5-R52H5) at 2 μg/mL (100 μL/well) can bind Anti-C5a (Human IgG1) with a linear range of 0.4-12.5 ng/mL (QC tested).
Catalog No. 50-195-6905 Supplier ACROBiosystems Supplier No. CO5R52H5100UG
Recombinant Protein;100UG;Rat Complement C5, His Tag (CO5-R52H5) is expressed from human 293 cells (HEK293). It contains AA Gln 24 - Ala 1688 (Accession # XP_001079130.2).;This protein carries a polyhistidine tag at the C-terminus.The mature form of Complement C5 is a disulfide-linked heterodimer composed of proteolytically cleaved α and β chain. Each α and β chain has a calculated MW of 74.1 kDa (β chain) and 114.8 kDa (α chain). The protein migrates as 70 kDa (β chain) and 110 kDa (α chain) under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Rat Complement C5, His Tag (Cat. No. CO5-R52H5) at 2 μg/mL (100 μL/well) can bind Anti-C5a (Human IgG1) with a linear range of 0.4-12.5 ng/mL (QC tested).