Learn More
ACROBiosystems ACROBiosystems Human Complement Component C2 Protein, His Tag
Catalog No. 501956493
Encompass_Preferred
Recombinant Protein;1MG;Human C2, His Tag (CO2-H52H7) is expressed from human 293 cells (HEK293). It contains AA Ala 21 - Leu 752 (Accession # P06681-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 83.0 kDa.
Catalog No. 50-195-6493 Supplier ACROBiosystems Supplier No. CO2H52H71MG
Recombinant Protein;1MG;Human C2, His Tag (CO2-H52H7) is expressed from human 293 cells (HEK293). It contains AA Ala 21 - Leu 752 (Accession # P06681-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 83.0 kDa.