Learn More
ACROBiosystems ACROBiosystems Human Complement Component C2 Protein, His Tag
Supplier:  ACROBiosystems CO2H52H750UG
Encompass_Preferred
Recombinant Protein;50UG;Human C2, His Tag (CO2-H52H7) is expressed from human 293 cells (HEK293). It contains AA Ala 21 - Leu 752 (Accession # P06681-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 83.0 kDa.
Catalog No. 50-195-6494