Learn More
ACROBiosystems ACROBiosystems Human Complement Component C5a Protein, Fc Tag
Catalog No. 501956752
Encompass_Preferred
Recombinant Protein;100UG;Human C5a, Fc Tag (C5A-H525a) is expressed from human 293 cells (HEK293). It contains AA Leu 679 - Arg 751 (Accession # P01031-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 34.6 kDa. The protein migrates as 38-40 kDa and 41-44 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human C5a, Fc Tag (Cat. No. C5A-H525a) at 2 μg/mL (100 μL/well) can bind Anti-C5a (Human IgG1) with a linear range of 2-16 ng/mL (QC tested).
Catalog No. 50-195-6752 Supplier ACROBiosystems Supplier No. C5AH525A100UG
Recombinant Protein;100UG;Human C5a, Fc Tag (C5A-H525a) is expressed from human 293 cells (HEK293). It contains AA Leu 679 - Arg 751 (Accession # P01031-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 34.6 kDa. The protein migrates as 38-40 kDa and 41-44 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human C5a, Fc Tag (Cat. No. C5A-H525a) at 2 μg/mL (100 μL/well) can bind Anti-C5a (Human IgG1) with a linear range of 2-16 ng/mL (QC tested).