Learn More
ACROBiosystems ACROBiosystems Mouse Complement Component C5a Protein, His Tag
Supplier:  ACROBiosystems C5AM51H4100UG
Encompass_Preferred
Recombinant Protein;100UG;Mouse C5a, His Tag (C5A-M51H4) is expressed from E.coli cells. It contains AA Asn 679 - Arg 755 (Accession # P06684).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 10.9 kDa. The protein migrates as 14 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Mouse C5a, His Tag (Cat. No. C5A-M51H4) at 2 μg/mL (100 μL/well) can bind Anti-C5a (Human IgG1) with a linear range of 0.4-6 ng/mL (QC tested).
Catalog No. 50-195-6974