Learn More
ACROBiosystems ACROBiosystems Cynomolgus Complement Component C5a Protein, Tag Free
Supplier:  ACROBiosystems C5AC51181MG
Encompass_Preferred
Recombinant Protein;1MG;Cynomolgus C5a, Tag Free (C5A-C5118) is expressed from E.coli cells. It contains AA Met 678 - Arg 751 (Accession # XP_015292262.1).;This protein carries no "tag". The protein has a calculated MW of 8.4 kDa. The protein migrates as 11 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Cynomolgus C5a, Tag Free (Cat. No. C5A-C5118) at 2 μg/mL (100 μL/well) can bind Anti-C5a (Human IgG1) with a linear range of 0.8-13 ng/mL (QC tested).
Catalog No. 50-195-6910