Learn More
ACROBiosystems ACROBiosystems Human Complement Component C5a Protein, Tag Free
Supplier:  ACROBiosystems C5AH511650UG
Encompass_Preferred
Recombinant Protein;50UG;Human C5a, Tag Free (C5A-H5116) is expressed from E.coli cells. It contains AA Leu 679 - Arg 751 (Accession # P01031).;This protein carries no "tag". The protein has a calculated MW of 8.3 kDa. The protein migrates as 9 kDa under reducing (R) condition (SDS-PAGE).;Immobilized Human C5a, Tag Free (Cat. No. C5A-H5116) at 2 μg/mL (100 μL/well) can bind Anti-C5a MAb, Human IgG1 with a linear range of 0.8-13 ng/mL (QC tested).;Human C5a, Tag Free (Cat. No. C5A-H5116) induce N-acetyl-β-D-glucosaminidase release from differentiated U937 cells. The ED50 for this effect is 0.215-0.323 μg/mL (Routinely tested).
Catalog No. 50-195-7484