Learn More
ACROBiosystems ACROBiosystems Rhesus macaque Complement Factor D / CFD Protein, Fc Tag
Catalog No. 501956814
Encompass_Preferred
Recombinant Protein;100UG;Rhesus macaque Complement Factor D, Fc Tag (CFD-R5255) is expressed from human 293 cells (HEK293) and inactive. It contains AA Ile 26 - Ala 253 (Accession # H9EXC1-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 51.0 kDa. The protein migrates as 53 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-195-6814 Supplier ACROBiosystems Supplier No. CFDR5255100UG
Recombinant Protein;100UG;Rhesus macaque Complement Factor D, Fc Tag (CFD-R5255) is expressed from human 293 cells (HEK293) and inactive. It contains AA Ile 26 - Ala 253 (Accession # H9EXC1-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 51.0 kDa. The protein migrates as 53 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.