Learn More
ACROBiosystems ACROBiosystems Human Complement Factor D / CFD Protein, Fc Tag
Catalog No. 501956817
Encompass_Preferred
Recombinant Protein;1MG;Human Complement Factor D, Fc Tag (CFD-H5256) is expressed from human 293 cells (HEK293) and inactive. It contains AA Ile 26 - Ala 253 (Accession # P00746-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 50.8 kDa. The protein migrates as 53 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-195-6817 Supplier ACROBiosystems Supplier No. CFDH52561MG
Recombinant Protein;1MG;Human Complement Factor D, Fc Tag (CFD-H5256) is expressed from human 293 cells (HEK293) and inactive. It contains AA Ile 26 - Ala 253 (Accession # P00746-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 50.8 kDa. The protein migrates as 53 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.