Learn More
ACROBiosystems ACROBiosystems Human Complement Factor D / CFD Protein, His Tag
Catalog No. 501957019
Encompass_Preferred
Recombinant Protein;1MG;Human Complement Factor D, His Tag (CFD-H52H8) is expressed from human 293 cells (HEK293) and active. It contains AA Ile 26 - Ala 253 (Accession # P00746-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 26.3 kDa. The protein migrates as 28 kDa under non-reducing (NR) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-195-7019 Supplier ACROBiosystems Supplier No. CFDH52H81MG
Recombinant Protein;1MG;Human Complement Factor D, His Tag (CFD-H52H8) is expressed from human 293 cells (HEK293) and active. It contains AA Ile 26 - Ala 253 (Accession # P00746-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 26.3 kDa. The protein migrates as 28 kDa under non-reducing (NR) condition (SDS-PAGE) due to glycosylation.