Learn More
ACROBiosystems ACROBiosystems Rhesus macaque / Cynomolgus Complement Factor D / CFD Protein, His Tag
Supplier:  ACROBiosystems CFDR52H31MG
Encompass_Preferred
Recombinant Protein;1MG;Rhesus macaque / Cynomolgus Complement Factor D, His Tag (CFD-R52H3) is expressed from human 293 cells (HEK293). It contains AA Ile 26 - Ala 253 (Accession # H9EXC1-1). In the region Ile 26 - Ala 253, the AA sequence of Rhesus macaque and Cynomolgus Complement Factor D are homologus.;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 26.5 kDa. The protein migrates as 25-27 kDa under non-reducing (NR) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-195-6996