Learn More
Chemglass Life Sciences COMPLETE Opti-Chem 15-Position Parallel Purification Tower
SDP
Supplier:  Chemglass Life Sciences OP6601102
Encompass_Preferred
COMPLETE Opti-Chem 15-Position Parallel Purification Tower. The Opti-Chem Personal Parallel Purification System puts purification and work-up in the hands of every synthetic chemist with fast and reliable results. Designed to match the requirements of drug discovery applications, a variety of Opti-Chem micro titer plate footprint collection blocks holding standard vials and tubes are available. These blocks are compatible with all centrifuge vacuum evaporators that use this common footprint, making going from purification to evaporation a snap. The Opti-Chem Personal Parallel Purification Tower is constructed from high quality, inert materials allowing all common solvents and buffers to be used with standard filtration, SPE, SS-LLE and silica packed columns. CG-6601-102 Consists of the Following: 2 each: OP-6601-11 Cartridge Holder, 15-Position, 3 packs: OP-6601-50 TFE Needles, 12/pack, 1 each: OP-6601-01 Base with Uprights
Catalog No. 50-153-2715