Learn More
Stoelting™ Complete Active Surgical Gas Anesthesia System
SDP
Catalog No. 10000937 Shop All Stoelting Co Products
Change view
Click to view available options
For Use With (Application):
Neuroscience
Catalog No. For Use With (Application)
10-000-937 Neuroscience
1 options
Catalog No. 10-000-937 Supplier Stoelting™ Supplier No. 53930
Add to Cart
Add to Cart
Request a Quote
Stoelting™ Complete Anesthesia System was designed specifically for stereotaxic surgery.
Complete system includes a tabletop and/or mobile cart with Isoflurane Vaporizer, Active Gas Scavenger and continuous flow Induction Chamber. Stereotaxic instrument sold separately.
Specifications
For Use With (Application) Neuroscience