Learn More
GSC Go Science Crazy Complete Cartesian Diver Kit
SCI_ED
Catalog No. S41122 Shop All GSC Products
Click to view available options
Includes:
38mm H Cartesian diver, 24 x 4cm hydrometer cylinder, Rubber diaphragm, instruction sheet with demonstration suggestions
Catalog No. Includes
S41122 38mm H Cartesian diver, 24 x 4cm hydrometer cylinder, Rubber diaphragm, instruction sheet with demonstration suggestions
1 options
Catalog No. S41122 Supplier GSC Go Science Crazy Supplier No. 3051
Includes
  • 38mm Cartesian diver
  • Hydrometer cylinder, 24 x 4cm
  • Rubber diaphragm
  • Instruction sheet with demonstration suggestions
Cartesian diver demonstrates Archimedes' principle of buoyancy.
  • Show the transmission of pressure by a liquid, compressibility of gases, and Archimedes' Principle
  • Hollow glass diver dips and returns to the surface of the cylinder using a small hole that admits air and water
  • Extra divers available separately
Specifications
For Use With (Application) Can be used as replacement to Cartesian Diver Unit or used in other cylinders or soda pop bottles experiments to demonstrates Archimedes' principle of buoyancy
Includes 38mm H Cartesian diver, 24 x 4cm hydrometer cylinder, Rubber diaphragm, instruction sheet with demonstration suggestions
Product Type Complete Cartesian Diver Apparatus
WARNING: Cancer - www.P65Warnings.ca.gov