Learn More
Quantimetrix 25-Oh Vitamin D2 & D3 Control, Complete D. Liquid, Made with Human Serum
SDP
Catalog No. 501528626
Encompass_Preferred
64-17-5. 25-OH Vitamin D2 & D3 Control, Complete D. Liquid, Made with Human Serum. Three-Year Refrigerated Shelf Life and 1-Year Open Vial Stability when Stored at 2Û8 Deg. C or 30 Days at Room Temperature.Two Level Set, 4 X3 mL.
This item is not returnable. View return policy
Catalog No. 50-152-8626 Supplier Quantimetrix Supplier No. 129001
Add to Cart
Add to Cart
64-17-5. 25-OH Vitamin D2 & D3 Control, Complete D. Liquid, Made with Human Serum. Three-Year Refrigerated Shelf Life and 1-Year Open Vial Stability when Stored at 2Û8 Deg. C or 30 Days at Room Temperature.Two Level Set, 4 X3 mL.