Learn More
MSC Complete Environmental Products Granular Absorbent, 2 cu ft, 1 ea
Catalog No. 5000796823
Encompass
The Kenaf Bio-Sorb granular absorbent features indigenous oil-eating microbes that address long-term spill disposal situations. This product is an all-natural, non-toxic, and fully biodegradable solution.
  • All-natural
  • Non-toxic
  • Completely biodegradable
  • Contains indigenous oil-eating microbes
Catalog No. 50-007-96823 Supplier MSC Supplier No. 09473901
The Kenaf Bio-Sorb granular absorbent features indigenous oil-eating microbes that address long-term spill disposal situations. This product is an all-natural, non-toxic, and fully biodegradable solution.
  • All-natural
  • Non-toxic
  • Completely biodegradable
  • Contains indigenous oil-eating microbes