Learn More
Complete Lever Outfit
SCI_ED
Catalog No. S40860
Change view
Click to view available options
Complete Lever Outfit
Catalog No.
S40860 Complete Lever Outfit
1 options
Catalog No. S40860
Includes
One meter rule, three knife-edge lever clamps, set of nine universal hooked masses from 10g to 1kg. Ring stand sold separately.
Learn how to apply leverage.
Set includes equipment for experiments on three classes of levers.