Learn More
ATHENA ES Complete Media Optimization Kit (Used by customers to identify the best Expression Medium for producing a recombinant protein.)
Catalog No. 5000158170
Encompass_Preferred
The Complete Media Optimization Kit™ contains 15 media formulations -- 10 of Athena's Proprietary Expression Media formulations along with 3 reference media and their APF counterparts. Since the production of a given recombinant protein can vary significantly depending on the type of medium employed, the MOK™ enables researchers to identify the best media formulation that yields the highest amount of target recombinant protein. Each dry powder kit contains enough mix to prepare 1 liter of each of the 15 media. Includes LB Miller, APF LB Miller, LB Lennox, APF LB Lennox, Hyper, Glucose M9Y, Turbo, Turbo Prime, Turbo Prime-olate, Superior, Superior Prime, Superior Prime-olate, Power, Power Prime, Power Prime-olate.
Catalog No. 50-001-58170 Supplier ATHENA ES Supplier No. 0185
The Complete Media Optimization Kit™ contains 15 media formulations -- 10 of Athena's Proprietary Expression Media formulations along with 3 reference media and their APF counterparts. Since the production of a given recombinant protein can vary significantly depending on the type of medium employed, the MOK™ enables researchers to identify the best media formulation that yields the highest amount of target recombinant protein. Each dry powder kit contains enough mix to prepare 1 liter of each of the 15 media. Includes LB Miller, APF LB Miller, LB Lennox, APF LB Lennox, Hyper, Glucose M9Y, Turbo, Turbo Prime, Turbo Prime-olate, Superior, Superior Prime, Superior Prime-olate, Power, Power Prime, Power Prime-olate.