Learn More
Sigma Aldrich Fine Chemicals Biosciences cOmplete, Mini, EDTA-free Protease Inhibitor Cocktail, 30 tablets
Catalog No. NC1921673
Encompass
cOmplete, Mini, EDTA-free, is used for the inhibition of serine and cysteine proteases in bacterial, yeast, plant, and animal cell extracts.
Catalog No. NC1921673 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 4693159001
cOmplete, Mini, EDTA-free, is used for the inhibition of serine and cysteine proteases in bacterial, yeast, plant, and animal cell extracts.