Learn More
Chemglass Life Sciences Complete Package, 0.2-0.5mL Microwave Vials, 8mm OD X 83mm Long (0.2 - 0.5mL Glass Vials require the PEEK adapter collar to attach crimp seals and septa. This is not included or sold by Chemglass.)
SDP
Catalog No. 501531783
Encompass_Preferred
Complete Package, 0.2-0.5mL Microwave Vials, 8mm OD X 83mm Long. 0.2 - 0.5mL Glass Vials require the PEEK adapter collar to attach crimp seals and septa. These are not included. Perform microwave-heated reactions in glass vials secured by 20mm aluminum seals with 0.125-inch thick, blue PTFE / white silicone septa. Glass vials are made from heavy-wall, Type I, Class A borosilicate for its high resistance to both chemicals and thermal shock. Microwave reaction vials are designed to withstand pressures up to 300 PSI (20 bars). PTFE-coated stir bars are used to provide continuous stirring and even heating during reactions.
Catalog No. 50-153-1783 Supplier Chemglass Life Sciences Supplier No. CG492004
Complete Package, 0.2-0.5mL Microwave Vials, 8mm OD X 83mm Long. 0.2 - 0.5mL Glass Vials require the PEEK adapter collar to attach crimp seals and septa. These are not included. Perform microwave-heated reactions in glass vials secured by 20mm aluminum seals with 0.125-inch thick, blue PTFE / white silicone septa. Glass vials are made from heavy-wall, Type I, Class A borosilicate for its high resistance to both chemicals and thermal shock. Microwave reaction vials are designed to withstand pressures up to 300 PSI (20 bars). PTFE-coated stir bars are used to provide continuous stirring and even heating during reactions.