Learn More
Bullard™ Complete Powered Air Purifying Respirator (PAPR) Kit
SDP
Catalog No. 19021491 Shop All Bullard Company Products
Change view
Click to view available options
EVAPAN4
Catalog No.
19-021-491 EVAPAN4
1 options
Catalog No. 19-021-491 Supplier Bullard™ Supplier No. EVAPAN4
Add to Cart
Add to Cart
NIOSH-approved PAPR provides protection and comfort alternatively to an N95 style mask. Ideal for healthcare professionals.
  • Provides healthcare workers with liquid protection
  • No fit testing is required for positive pressure PAPR
  • Exhaled air filtration for any critical environment or sterile field applications
  • Two speed motor blower offers either 7.0 or 8.5 CFM (Depending on operator's preference)
  • Unique hood is constructed of Tychem™ 2000 to meet ASTM-1671 requirements
  • HEPA filtration panel on bottom part of hood
  • Alarms to alert of low airflow and low battery provide additional piece of mind during operation
  • Visual fuel gauge allows healthcare workers to approximate battery run time, which can last up to 12 hours on single charge
  • Lithium polymer batteries are easy to maintain, charging/memory issues are often not associated with NiCad batteries
  • Entire assembly can be wiped down with a variety of disinfectants
  • Closed cell foam comfort belt is easy to clean and maximizes ergonomic fit
Specifications
Assigned Protection Factor (APF) 25
Battery Life 10 to 12 hours
Battery Type Lithium Polymer
Belt Material Closed Cell Foam
Breathing Tube Length 32 in.
Product Type PAPR Healthcare Systems and Kits
Includes Motor Blower Unit, Air Flow Indicator, Battery, Charger, 20 LF2LHE Hood, PAHBT Breathing Tube, and PAPRFC3 HEPA Filter
Model EVAPAN4
Recharge Time 4 Hr.
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.