Learn More
Sigma Aldrich Fine Chemicals Biosciences cOmplete Protease Inhibitor Cocktail, 3 x 20 tablets
Catalog No. NC0930105
Encompass
cOmplete Protease Inhibitor Tablets inhibit a broad spectrum of serine, cysteine, and metalloproteases, as well as calpains. Due to the optimized composition of the tablets, they show excellent inhibition effects, and are well suited for the protection of proteins isolated from animal tissues, plants, yeast, and bacteria. cOmplete tablets contain EDTA and both irreversible and reversible protease inhibitors.
Catalog No. NC0930105 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 11836145001
Add to Cart
Add to Cart
cOmplete Protease Inhibitor Tablets inhibit a broad spectrum of serine, cysteine, and metalloproteases, as well as calpains. Due to the optimized composition of the tablets, they show excellent inhibition effects, and are well suited for the protection of proteins isolated from animal tissues, plants, yeast, and bacteria. cOmplete tablets contain EDTA and both irreversible and reversible protease inhibitors.