Learn More
Chemglass Life Sciences 30L Process Reactor, Complete, Electric Motor, 82.6" OAH with Motor, 18.6" Clearance Below Valve
SDP
Catalog No. CG1968630
Encompass_Preferred
30L Process Reactor, Complete, Electric Motor, 82.6" OAH with Motor, 18.6" Clearance Below Valve.¿ Motor Mount Arm Can Unlock & Swing Out Giving Greater Access To Reactor¿ Digital Temperature Monitor¿ Electric And XP Motor Has Separate Controller With Digital Display¿ Smaller Footprint Support Frame With Oversized Rugged S.S.Uprights And Heavy Duty PTFE Coated Aluminum Fittings¿ Reactor Jacket Graduation Scale On 2 Sides¿ Lower Safety Ring¿ Zero Dead Space Detachable Drain Valve Has Knob With Laser Etched Arrows For Open And Close¿ Electric Or Air Motor Optional XP Motor Available¿ Open Frame Design For Excellent Accessibility¿ 7-Neck Lid With Large Powder Addition Port¿ Support Plate Holds Reactor Securely & Allows Easy Installation/Removal Of Reactor ¿ Ridge Clamp Accepts 1/2¿ Rod And Adds Additional Support For Large Glassware¿ S.S.Inlet/Outlet Adapters Available In M16 X 1 And M30 X 1.5 Allow Connection To Cooling/Heating Circulator¿ Heavy Duty Lockable Casters
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Catalog No. CG1968630 Supplier Chemglass Life Sciences Supplier No. CG1968630
Add to Cart
Add to Cart
Request a Quote
30L Process Reactor, Complete, Electric Motor, 82.6" OAH with Motor, 18.6" Clearance Below Valve.¿ Motor Mount Arm Can Unlock & Swing Out Giving Greater Access To Reactor¿ Digital Temperature Monitor¿ Electric And XP Motor Has Separate Controller With Digital Display¿ Smaller Footprint Support Frame With Oversized Rugged S.S.Uprights And Heavy Duty PTFE Coated Aluminum Fittings¿ Reactor Jacket Graduation Scale On 2 Sides¿ Lower Safety Ring¿ Zero Dead Space Detachable Drain Valve Has Knob With Laser Etched Arrows For Open And Close¿ Electric Or Air Motor Optional XP Motor Available¿ Open Frame Design For Excellent Accessibility¿ 7-Neck Lid With Large Powder Addition Port¿ Support Plate Holds Reactor Securely & Allows Easy Installation/Removal Of Reactor ¿ Ridge Clamp Accepts 1/2¿ Rod And Adds Additional Support For Large Glassware¿ S.S.Inlet/Outlet Adapters Available In M16 X 1 And M30 X 1.5 Allow Connection To Cooling/Heating Circulator¿ Heavy Duty Lockable Casters
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.