Learn More
Chemglass Life Sciences 50L Process Reactor, Complete, Air Motor, 87.3" OAH with Motor, 16.2" Clearance Below Valve
SDP
Catalog No. CG1970650
Encompass_Preferred
50L Process Reactor, Complete, Air Motor, 87.3" OAH with Motor, 16.2" Clearance Below Valve. Features:¿ Motor Mount Arm Can Unlock And Swing Out Giving Greater Access To Reactor¿ Digital Temperature Monitor¿ Electric And XP Motor Has Separate Controller With Digital Display¿ Smaller Footprint Support Frame With Oversized Rugged S.S. Uprights And Heavy Duty PTFE Coated Aluminum Fittings¿ Reactor Jacket Graduation Scale On Both Sides¿ Lower Safety Ring¿ Zero Dead Space Detachable Drain Valve Has Knob With Laser Etched Directional Arrows For Open And Close¿ Electric Or Air Motor Optional XP Motor Available¿ Open Frame Design Provides Excellent Accessibility¿ 7-Neck Lid With Large Powder Addition Port¿ Reactor Support Plate Holds Reactor Securely¿ Ridge Clamp Accepts 1/2¿ Rod And Adds Additional Support For Large Glassware¿ S.S. Inlet/Outlet Adapters Available In M16 X 1 And M30 X 1.5 Allow Connection To Cooling/Heating Circulator¿ Heavy Duty Lockable Casters
Catalog No. CG1970650 Supplier Chemglass Life Sciences Supplier No. CG1970650
Add to Cart
Add to Cart
Request a Quote
50L Process Reactor, Complete, Air Motor, 87.3" OAH with Motor, 16.2" Clearance Below Valve. Features:¿ Motor Mount Arm Can Unlock And Swing Out Giving Greater Access To Reactor¿ Digital Temperature Monitor¿ Electric And XP Motor Has Separate Controller With Digital Display¿ Smaller Footprint Support Frame With Oversized Rugged S.S. Uprights And Heavy Duty PTFE Coated Aluminum Fittings¿ Reactor Jacket Graduation Scale On Both Sides¿ Lower Safety Ring¿ Zero Dead Space Detachable Drain Valve Has Knob With Laser Etched Directional Arrows For Open And Close¿ Electric Or Air Motor Optional XP Motor Available¿ Open Frame Design Provides Excellent Accessibility¿ 7-Neck Lid With Large Powder Addition Port¿ Reactor Support Plate Holds Reactor Securely¿ Ridge Clamp Accepts 1/2¿ Rod And Adds Additional Support For Large Glassware¿ S.S. Inlet/Outlet Adapters Available In M16 X 1 And M30 X 1.5 Allow Connection To Cooling/Heating Circulator¿ Heavy Duty Lockable Casters