Learn More
MOLTOX Molecular Toxicology Inc Salmonella Mutagenicity Test Kit
SDP
Supplier:  MOLTOX Molecular Toxicology Inc 31100.2
Encompass
The materials contained in this Salmonella Mutagenicity Assay Kit include virtually all of the supplies necessary for the conduct of the “Ames Assay” as described by Maron & Ames (Maron, D. M. and B. N. Ames, Revised methods for the Salmonella mutagenicity test, Mutation Research, 113: 173-215, 1983) and Mortelmans & Zeiger (Mortelmans, K. and E. Zeiger, The Ames Salmonella/microsome mutagenicity assay, Mutation Research, 455: 29-60, 2000). We strongly recommend that you carefully read one of these papers and OECD guideline 471 before you attempt to perform the assay. This kit contains four tester strains; TA1535, TA1537, TA98, and TA100. Tester strains TA98 and TA100 carry a plasmid (pKM101); TA1535 and TA1537 contain no plasmids. The MOLTOX 31-100.2 kit includes sufficient components for testing 1 unknown sample, in triplicate analysis of 5 concentrations, positive and negative controls, with and without S9.
Catalog No. 50-948-801