Learn More
Electron Microscopy Sciences Lyophilization Complete Tear Off Seals, 7 mm I.D., 13 mm O.D.
SDP
Catalog No. 5032786
Encompass_Preferred
Aluminum lyophilization seals are unlined. Seal can be completely removed. Fits mouth ID 7mm x OD 13mm.
Catalog No. 50-327-86 Supplier Electron Microscopy Sciences Supplier No. 6032207
Aluminum lyophilization seals are unlined. Seal can be completely removed. Fits mouth ID 7mm x OD 13mm.