Learn More
MSC Home Safeguard Test Kit, Heat, Smoke and CO Detector, 1 ea
Catalog No. 5000685542
Encompass
This comprehensive test kit provides a complete solution for the maintenance and verification of smoke, heat, and carbon monoxide detectors. Designed to facilitate compliance with NFPA 72 and International Fire Code standards, the system allows for testing without the use of ladders or unsafe climbing.
  • Includes enclosed delivery test kit, open delivery test kit, and chemical heat detector tester
  • Enables testing of smoke, heat, and CO detectors
  • Utilizes durable, lightweight, and non-conductive fiberglass extension poles
  • Features a versatile dispenser compatible with multiple aerosol can sizes
  • Supports safe operation by eliminating the need to climb or overreach
Catalog No. 50-006-85542 Supplier MSC Supplier No. 63435341
Add to Cart
Add to Cart
This comprehensive test kit provides a complete solution for the maintenance and verification of smoke, heat, and carbon monoxide detectors. Designed to facilitate compliance with NFPA 72 and International Fire Code standards, the system allows for testing without the use of ladders or unsafe climbing.
  • Includes enclosed delivery test kit, open delivery test kit, and chemical heat detector tester
  • Enables testing of smoke, heat, and CO detectors
  • Utilizes durable, lightweight, and non-conductive fiberglass extension poles
  • Features a versatile dispenser compatible with multiple aerosol can sizes
  • Supports safe operation by eliminating the need to climb or overreach