Learn More
Chemglass Life Sciences Vacuum Trap, Complete, Trap Bottle 45mm OD x 250mm Overall Length, 16mm OD Side and Inner Tubes, 40/50 Jt.
SDP
Catalog No. CG451407
Encompass_Preferred
Vacuum Trap, Complete, Trap Bottle 45mm OD x 250mm Overall Length, 16mm OD Side and Inner Tubes, 40/50 Jt. Two piece vacuum trap connected with an inner and outer joint to facilitate cleaning.
Catalog No. CG451407 Supplier Chemglass Life Sciences Supplier No. CG451407
Vacuum Trap, Complete, Trap Bottle 45mm OD x 250mm Overall Length, 16mm OD Side and Inner Tubes, 40/50 Jt. Two piece vacuum trap connected with an inner and outer joint to facilitate cleaning.