Learn More
PATHOLOGY INNOVATIONS Comes with 3 well bars (18mm, 24mm, 30mm), 6 sm. chucks, 4 lg. chucks, 4 over-chuck freezing blocks, chuck bin, 16 dispensing slides, cutting board/freezing griddle, elevated freezing block, and accessories. Forceps not included.
Catalog No. NC3370378
Encompass_Preferred
The Peters Precision Cryoembedding System is a combination of simple techniques and apparatus which allows the user to embed tissues for frozen section with a level of control and predictability that can surpass even paraffin embedding. The Complete Well Bar Set comes with 3 well bars (18mm, 24mm, 30mm), 6 sm. chucks, 4 lg. chucks, 4 over-chuck freezing blocks, chuck bin, 16 dispensing slides, cutting board/freezing griddle, elevated freezing block, and accessories. Forceps not included. This set is made for outfitting labs that process a high volume of frozen sections with various sized speciments, and a great way to get started using the Peters Precision Cryoembedding System which has been adopted by thousands of hospitals globally. Learn how to use the system as well as advanced frozen section technique at pathologyinnovations.com.
Catalog No. NC3370378 Supplier PATHOLOGY INNOVATIONS Supplier No. 101
The Peters Precision Cryoembedding System is a combination of simple techniques and apparatus which allows the user to embed tissues for frozen section with a level of control and predictability that can surpass even paraffin embedding. The Complete Well Bar Set comes with 3 well bars (18mm, 24mm, 30mm), 6 sm. chucks, 4 lg. chucks, 4 over-chuck freezing blocks, chuck bin, 16 dispensing slides, cutting board/freezing griddle, elevated freezing block, and accessories. Forceps not included. This set is made for outfitting labs that process a high volume of frozen sections with various sized speciments, and a great way to get started using the Peters Precision Cryoembedding System which has been adopted by thousands of hospitals globally. Learn how to use the system as well as advanced frozen section technique at pathologyinnovations.com.